ENA018104328 Enamine

4-(4-Chloro-phenylsulfanyl)-butyric acid

  • MDL number MFCD00460787
  • Cas Number : 29193-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00233

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00233
Availability details for
Quantity
  • Availability
  • Add to Cart