ENA018104330 Enamine

2,3,6-trichlorobenzoic acid

  • MDL number MFCD00053310
  • Cas Number : 50-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00235

Safety Information

signalword Warning
pcodes P264,P270,P273,P301+P312,P330,P391,P501
hcodes Xn,N
MSDS https://www.enaminestore.com/catalog/EN300-00235
s_phrases_eu 61
pictograms GHS07,GHS09
hstatements H302,H411
Availability details for
Quantity
  • Availability
  • Add to Cart