ENA018104333 Enamine

3-Benzothiazol-2-yl-4-(4-methoxy-phenyl)-but-3-enoic acid

  • MDL number MFCD03654407
  • Cas Number : 748786-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00238

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00238
Availability details for
Quantity
  • Availability
  • Add to Cart