ENA018104337 Enamine

3-Benzothiazol-2-yl-4-(3,4-dimethoxy-phenyl)-but-3-enoic acid

  • MDL number MFCD03654409
  • Cas Number : 748786-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00242
Availability details for
Quantity
  • Availability
  • Add to Cart