ENA018104342 Enamine

4-{2,4-dioxo-3-azatricyclo[7.3.1.0,5,13]trideca-1(13),5,7,9,11-pentaen-3-yl}butanoic acid

  • MDL number MFCD00181400
  • Cas Number : 88909-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-87
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00247
Availability details for
Quantity
  • Availability
  • Add to Cart