ENA018104357 Enamine

4-(4-chlorophenyl)-2-oxo-3-(quinolin-2-yl)but-3-enoic acid

  • MDL number MFCD03657977
  • Cas Number : 473396-84-8
  • Molecular Weight : 337.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00262
Availability details for
Quantity
  • Availability
  • Add to Cart