ENA018104373 Enamine

3-(Morpholine-4-sulfonyl)-benzoic acid

  • MDL number MFCD00622729
  • Cas Number : 299181-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00278
Availability details for
Quantity
  • Availability
  • Add to Cart