ENA018104375 Enamine

3-(benzylsulfamoyl)benzoic acid

  • MDL number MFCD00622724
  • Cas Number : 7326-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00280
Availability details for
Quantity
  • Availability
  • Add to Cart