ENA018104377 Enamine

2-hydroxy-5-(thiophene-2-sulfonamido)benzoic acid

  • MDL number MFCD02700612
  • Cas Number : 326023-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00282
Availability details for
Quantity
  • Availability
  • Add to Cart