ENA018104398 Enamine

3-(3,4-Dihydro-1H-isoquinoline-2-sulfonyl)-benzoic acid

  • MDL number MFCD01197550
  • Cas Number : 327092-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...58
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00303

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00303
Availability details for
Quantity
  • Availability
  • Add to Cart