ENA018104400 Enamine

3-{[3-(trifluoromethyl)phenyl]sulfamoyl}benzoic acid

  • MDL number MFCD02708228
  • Cas Number : 327092-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00305
Availability details for
Quantity
  • Availability
  • Add to Cart