ENA018104406 Enamine

4-[(4-methylphenyl)sulfamoyl]benzoic acid

  • MDL number MFCD03147233
  • Cas Number : 379254-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00311
Availability details for
Quantity
  • Availability
  • Add to Cart