ENA018104408 Enamine

4-(4-ethoxybenzenesulfonamido)benzoic acid

  • MDL number MFCD02704402
  • Cas Number : 326916-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00313
Availability details for
Quantity
  • Availability
  • Add to Cart