ENA018104421 Enamine

2-(4-bromobenzenesulfonamido)benzoic acid

  • MDL number MFCD00583502
  • Cas Number : 51012-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-175
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00326
Availability details for
Quantity
  • Availability
  • Add to Cart