ENA018104437 Enamine

4-chloro-3-[(4-chlorophenyl)sulfamoyl]benzoic acid

  • MDL number MFCD01122312
  • Cas Number : 325724-62-7
  • Molecular Weight : 346.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00342
Availability details for
Quantity
  • Availability
  • Add to Cart