ENA018104442 Enamine

5-Diethylsulfamoyl-2-hydroxy-benzoic acid

  • MDL number MFCD02697463
  • Cas Number : 66306-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00347
Availability details for
Quantity
  • Availability
  • Add to Cart