ENA018104450 Enamine

2,4-dichloro-5-(morpholine-4-sulfonyl)benzoic acid

  • MDL number MFCD00739271
  • Cas Number : 292644-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-215
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00355
Availability details for
Quantity
  • Availability
  • Add to Cart