ENA018104451 Enamine

4-bromo-3-(morpholine-4-sulfonyl)benzoic acid

  • MDL number MFCD00625739
  • Cas Number : 300383-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00356
Availability details for
Quantity
  • Availability
  • Add to Cart