ENA018104484 Enamine

4-[(2,6-dimethylmorpholin-4-yl)sulfonyl]benzoic acid

  • MDL number MFCD02705286
  • Cas Number : 328038-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00389
Availability details for
Quantity
  • Availability
  • Add to Cart