ENA018104490 Enamine

4-(Morpholine-4-sulfonyl)-benzoic acid

  • MDL number MFCD00441966
  • Cas Number : 10252-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 287...289
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00395
Availability details for
Quantity
  • Availability
  • Add to Cart