ENA018104497 Enamine

2-chloro-5-(piperidine-1-sulfonyl)benzoic acid

  • MDL number MFCD00625735
  • Cas Number : 109029-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00402
Availability details for
Quantity
  • Availability
  • Add to Cart