ENA018104502 Enamine

2-(3,4-dimethoxybenzenesulfonamido)benzoic acid

  • MDL number MFCD02711601
  • Cas Number : 325721-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00407
Availability details for
Quantity
  • Availability
  • Add to Cart