ENA018104512 Enamine

5-[(4-ethoxyphenyl)(methyl)sulfamoyl]-2-methylbenzoic acid

  • MDL number MFCD03147296
  • Cas Number : 379254-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00417
Availability details for
Quantity
  • Availability
  • Add to Cart