ENA018104513 Enamine

2-chloro-5-[(4-phenylpiperazin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD02720433
  • Cas Number : 380193-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00418
Availability details for
Quantity
  • Availability
  • Add to Cart