ENA018104516 Enamine

2-[5-(morpholine-4-sulfonyl)thiophen-2-yl]acetic acid

  • MDL number MFCD01820668
  • Cas Number : 309282-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00421
Availability details for
Quantity
  • Availability
  • Add to Cart