ENA018104517 Enamine

3-(2,3-Dihydro-indole-1-sulfonyl)-benzoic acid

  • MDL number MFCD02708219
  • Cas Number : 327092-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00422

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00422
Availability details for
Quantity
  • Availability
  • Add to Cart