ENA018104540 Enamine

4-Fluoro-3-(piperidine-1-sulfonyl)-benzoic acid

  • MDL number MFCD01197508
  • Cas Number : 311785-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00445
Availability details for
Quantity
  • Availability
  • Add to Cart