ENA018104541 Enamine

2-(4-fluoro-2-methylbenzenesulfonamido)benzoic acid

  • MDL number MFCD02708878
  • Cas Number : 327105-31-7
  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00446
Availability details for
Quantity
  • Availability
  • Add to Cart