ENA018104554 Enamine

3-[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)sulfamoyl]benzoic acid

  • MDL number MFCD01917220
  • Cas Number : 307511-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00459
Availability details for
Quantity
  • Availability
  • Add to Cart