ENA018104570 Enamine

2-fluoro-5-(morpholine-4-sulfonyl)benzoic acid

  • MDL number MFCD02699875
  • Cas Number : 326609-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00475
Availability details for
Quantity
  • Availability
  • Add to Cart