ENA018104603 Enamine

5-[(2-methoxyphenyl)(methyl)sulfamoyl]-2-methylbenzoic acid

  • MDL number MFCD02725171
  • Cas Number : 380342-16-5
  • Molecular Weight : 335.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00508
Availability details for
Quantity
  • Availability
  • Add to Cart