ENA018104611 Enamine

3,4-dichloro-5-[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)sulfamoyl]benzoic acid

  • MDL number MFCD03150312
  • Cas Number : 380349-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00517
Availability details for
Quantity
  • Availability
  • Add to Cart