ENA018104641 Enamine

2-[4-(N-methyl4-methylbenzenesulfonamido)phenoxy]acetic acid

  • MDL number MFCD03957574
  • Cas Number : 722467-62-1
  • Molecular Weight : 335.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 125-126
Iupac Name (4-{methyl[(4-methylphenyl)sulfonyl]amino}phenoxy)acetic acid
Inchi Code 1S/C16H17NO5S/c1-12-3-9-15(10-4-12)23(20,21)17(2)13-5-7-14(8-6-13)22-11-16(18)19/h3-10H,11H2,1-2H3,(H,18,19)
Inchi Key LGCHJSYHCNININ-UHFFFAOYSA-N

Description

Melting Point 125-126
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00548
Availability details for
Quantity
  • Availability
  • Add to Cart