ENA018104642 Enamine

2-[4-(N-methyl4-chlorobenzenesulfonamido)phenoxy]acetic acid

  • MDL number MFCD02724575
  • Cas Number : 380342-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00549
Availability details for
Quantity
  • Availability
  • Add to Cart