ENA018104646 Enamine

2-(4-Ethoxy-benzenesulfonyl)-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid

  • MDL number MFCD04971540
  • Cas Number : 1008091-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00554
Availability details for
Quantity
  • Availability
  • Add to Cart