ENA018104647 Enamine

2-(thiophene-2-sulfonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD03724567
  • Cas Number : 1008267-14-8
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00555
Availability details for
Quantity
  • Availability
  • Add to Cart