ENA018104661 Enamine

3-[ethyl(phenyl)sulfamoyl]-4-methylbenzoic acid

  • MDL number MFCD03152471
  • Cas Number : 379726-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00570
Availability details for
Quantity
  • Availability
  • Add to Cart