ENA018104685 Enamine

3-[Allyl-(2-methoxy-phenyl)-sulfamoyl]-benzoic acid

  • MDL number MFCD03478289
  • Cas Number : 438029-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00597
Availability details for
Quantity
  • Availability
  • Add to Cart