ENA018104688 Enamine

2-hydroxy-6-(morpholine-4-sulfonyl)quinoline-4-carboxylic acid

  • MDL number MFCD03150430
  • Cas Number : 380426-68-6
  • Molecular Weight : 338.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00600
Availability details for
Quantity
  • Availability
  • Add to Cart