ENA018104692 Enamine

2-(4-Ethoxy-benzenesulfonylamino)-3-(1H-indol-3-yl)-propionic acid

  • MDL number MFCD02724157
  • Cas Number : 1396966-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00604

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00604
Availability details for
Quantity
  • Availability
  • Add to Cart