ENA018104698 Enamine

4-[(3,4-dimethylphenyl)sulfamoyl]benzoic acid

  • MDL number MFCD03476378
  • Cas Number : 380437-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235-236
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00612
Availability details for
Quantity
  • Availability
  • Add to Cart