ENA018104699 Enamine

4-chloro-3-[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)sulfamoyl]benzoic acid

  • MDL number MFCD02711519
  • Cas Number : 109030-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00613
Availability details for
Quantity
  • Availability
  • Add to Cart