ENA018104709 Enamine

2-amino-5-[(2-methoxyphenyl)sulfamoyl]benzoic acid

  • MDL number MFCD03480279
  • Cas Number : 519153-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00623
Availability details for
Quantity
  • Availability
  • Add to Cart