ENA018104713 Enamine

4-(4-fluorobenzenesulfonamido)benzoic acid

  • MDL number MFCD00598811
  • Cas Number : 392313-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00627
Availability details for
Quantity
  • Availability
  • Add to Cart