ENA018104730 Enamine

4-Methoxy-3-[methyl-(2-pyridin-3-yl-ethyl)-sulfamoyl]-benzoic acid

  • MDL number MFCD03966885
  • Cas Number : 554405-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00645

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00645
Availability details for
Quantity
  • Availability
  • Add to Cart