ENA018104732 Enamine

3-[(3,5-dimethoxyphenyl)sulfamoyl]-4-methoxybenzoic acid

  • MDL number MFCD03655868
  • Cas Number : 710989-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235-236
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00647
Availability details for
Quantity
  • Availability
  • Add to Cart