ENA018104737 Enamine

3-(4-fluorobenzenesulfonamido)-4-methylbenzoic acid

  • MDL number MFCD03949337
  • Cas Number : 721406-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00652
Availability details for
Quantity
  • Availability
  • Add to Cart