ENA018104744 Enamine

4-methoxy-3-{[2-(pyridin-2-yl)ethyl]sulfamoyl}benzoic acid

  • MDL number MFCD03966914
  • Cas Number : 565166-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00659
Availability details for
Quantity
  • Availability
  • Add to Cart