ENA018104746 Enamine

4-methoxy-3-{[4-(pyridin-2-yl)piperazin-1-yl]sulfonyl}benzoic acid

  • MDL number MFCD03653046
  • Cas Number : 748776-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00661
Availability details for
Quantity
  • Availability
  • Add to Cart