ENA018104747 Enamine

2,4-dichloro-5-[(6-ethoxy-1,3-benzothiazol-2-yl)sulfamoyl]benzoic acid

  • MDL number MFCD03653042
  • Cas Number : 692745-62-3
  • Molecular Weight : 447.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-164
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00662
Availability details for
Quantity
  • Availability
  • Add to Cart